ENAH11E08A49 Enamine

methyl 2-hydroxy-2-(naphthalen-1-yl)acetate

  • MDL number MFCD16723798
  • Cas Number : 78772-53-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1869144

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1869144
Availability details for
Quantity
  • Availability
  • Add to Cart