ENAH11E08E8E Enamine

2-phenylbenzene-1-thiol

  • MDL number MFCD03840407
  • Cas Number : 2688-96-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 38-39
Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1870247

Safety Information

signalword Danger
pcodes P264,P270,P273,P280,P301+P312,P305+P351+P338,P310,P330,P391,P501
hcodes N,Xi,Xn
MSDS https://www.enaminestore.com/catalog/EN300-1870247
pictograms GHS05,GHS07,GHS09
hstatements H302,H318,H410
Availability details for
Quantity
  • Availability
  • Add to Cart