ENAH11E145D0 Enamine

1-(2-bromo-6-methoxyphenyl)ethan-1-one

  • MDL number MFCD22573041
  • Cas Number : 380225-68-3
  • Molecular Weight : 229.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1896836

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1896836
Availability details for
Quantity
  • Availability
  • Add to Cart