ENAH11E1AABA Enamine

1-(4-bromo-3-methoxyphenyl)ethan-1-one

  • MDL number MFCD17169058
  • Cas Number : 50870-44-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1912067

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1912067
Availability details for
Quantity
  • Availability
  • Add to Cart