ENAH11E1B652 Enamine

2-bromo-5-ethynylthiophene

  • MDL number MFCD08703509
  • Cas Number : 105995-73-1
  • Molecular Weight : 187.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1914636

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1914636
Availability details for
Quantity
  • Availability
  • Add to Cart