ENAH11E30AA6 Enamine

5-chloro-2-methoxypyridine-4-carboxylic acid

  • MDL number MFCD07375110
  • Cas Number : 88912-22-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1966390

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1966390
Availability details for
Quantity
  • Availability
  • Add to Cart