ENAH11E354CD Enamine

(1S)-1-(2-chloropyridin-3-yl)ethan-1-ol

  • MDL number MFCD32065867
  • Cas Number : 2227641-27-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1976810

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1976810
Availability details for
Quantity
  • Availability
  • Add to Cart