ENAH11E373F3 Enamine

3-(chloromethyl)-3-methyloxetane

  • MDL number MFCD01709552
  • Cas Number : 822-48-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1982101

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1982101
Availability details for
Quantity
  • Availability
  • Add to Cart