ENAH11E37A08 Enamine

5-(chloromethyl)-2-(propan-2-yl)-1,3-thiazole

  • MDL number MFCD06797200
  • Cas Number : 1051463-88-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1983475

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1983475
Availability details for
Quantity
  • Availability
  • Add to Cart