ENAH11E38E99 Enamine

3-(2-aminoethoxy)-1lambda6-thiolane-1,1-dione

  • MDL number MFCD16713541
  • Cas Number : 857626-00-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1987384

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1987384
Availability details for
Quantity
  • Availability
  • Add to Cart