ENAH11E4062D Enamine

9-chloro-3,4-dihydro-2H-1,5-benzodioxepine-7-sulfonyl chloride

  • MDL number MFCD31666477
  • Cas Number : 2228322-29-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2005124

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2005124
Availability details for
Quantity
  • Availability
  • Add to Cart