ENAH11E4088A Enamine

1-(3-methanesulfonylphenyl)cyclopentane-1-carboxylic acid

  • MDL number MFCD31657879
  • Cas Number : 1891969-16-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2004845

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2004845
Availability details for
Quantity
  • Availability
  • Add to Cart