ENAH11E40C02 Enamine

methyl 2-(3,3-difluorocyclobutyl)-2-hydroxyacetate

  • MDL number MFCD31718434
  • Cas Number : 2228290-05-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2005867

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2005867
Availability details for
Quantity
  • Availability
  • Add to Cart