ENAH11E4142F Enamine

methyl 1-amino-3-(difluoromethyl)cyclobutane-1-carboxylate hydrochloride

  • MDL number MFCD31666674
  • Cas Number : 2228157-61-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2007644

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2007644
Availability details for
Quantity
  • Availability
  • Add to Cart