ENAH11E41434 Enamine

methyl 2-amino-2-(1-methoxycyclobutyl)acetate hydrochloride

  • MDL number MFCD31666683
  • Cas Number : 2228084-18-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2007736

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2007736
Availability details for
Quantity
  • Availability
  • Add to Cart