ENAH11E41435 Enamine

sodium 2-[2-(oxolan-3-yl)-1,3-thiazol-4-yl]acetate

  • MDL number MFCD31666499
  • Cas Number : 2228777-36-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2007740

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2007740
Availability details for
Quantity
  • Availability
  • Add to Cart