ENAH11E41437 Enamine

6-(cyclobutylmethoxy)-1,2,3,4-tetrahydroisoquinoline; trifluoroacetic acid

  • MDL number MFCD31666638
  • Cas Number : 2228650-22-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data CF3COOH
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2007749

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2007749
Availability details for
Quantity
  • Availability
  • Add to Cart