ENAH11E41438 Enamine

tert-butyl (2S)-2-(5-amino-1-methyl-1H-1,2,4-triazol-3-yl)pyrrolidine-1-carboxylate

  • MDL number MFCD31666623
  • Cas Number : 2227781-93-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2007753

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2007753
Availability details for
Quantity
  • Availability
  • Add to Cart