ENAH11E4143B Enamine

3-[(tert-butyldimethylsilyl)oxy]-2-chloro-6-(methylsulfanyl)benzaldehyde

  • MDL number MFCD31666618
  • Cas Number : 2228154-44-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2007881

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2007881
Availability details for
Quantity
  • Availability
  • Add to Cart