ENAH11E4143C Enamine

ethyl 1-(iodomethyl)-2-oxabicyclo[2.1.1]hexane-4-carboxylate

  • MDL number MFCD31666502
  • Cas Number : 2229432-89-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2007889

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2007889
Availability details for
Quantity
  • Availability
  • Add to Cart