ENAH11E4143E Enamine

potassium 4'-(aminomethyl)-1-[(tert-butoxy)carbonyl]-3'-oxaspiro[azetidine-3,2'-bicyclo[2.1.1]hexane]-1'-carboxylate

  • MDL number MFCD31666600

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data K+
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2007918

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2007918
Availability details for
Quantity
  • Availability
  • Add to Cart