ENAH11E4144E Enamine

2-(1,3-dioxaindan-5-yl)-8-chloroimidazo[1,2-a]pyridin-3-amine hydrochloride

  • MDL number MFCD31690828
  • Cas Number : 2228908-11-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008124

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008124
Availability details for
Quantity
  • Availability
  • Add to Cart