ENAH11E4144F Enamine

2-methyl-2-azabicyclo[2.1.1]hexane-1-carboxylic acid hydrochloride

  • MDL number MFCD31666781
  • Cas Number : 2228313-13-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008131

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008131
Availability details for
Quantity
  • Availability
  • Add to Cart