ENAH11E41454 Enamine

tert-butyl (4S)-2-cyano-4-fluoropyrrolidine-1-carboxylate

  • MDL number MFCD31666741
  • Cas Number : 2227983-10-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008169

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008169
Availability details for
Quantity
  • Availability
  • Add to Cart