ENAH11E4145A Enamine

2-[(4-cyano-2-fluorophenyl)(methyl)amino]acetic acid hydrochloride

  • MDL number MFCD31666797
  • Cas Number : 2229137-64-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008454

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008454
Availability details for
Quantity
  • Availability
  • Add to Cart