ENAH11E4145C Enamine

4,6-dibromo-5-chloropyrimidine

  • MDL number MFCD31666645
  • Cas Number : 2229563-91-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008500

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008500
Availability details for
Quantity
  • Availability
  • Add to Cart