ENAH11E4145E Enamine

7-bromo-1H,4H,5H-imidazo[4,5-c]pyridin-4-one hydrochloride

  • MDL number MFCD31690736
  • Cas Number : 2229265-94-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008580

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008580
Availability details for
Quantity
  • Availability
  • Add to Cart