ENAH11E41460 Enamine

7,7-difluoro-2-azaspiro[3.5]nonane; trifluoroacetic acid

  • MDL number MFCD31666561
  • Cas Number : 2228781-03-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data CF3COOH
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008646

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008646
Availability details for
Quantity
  • Availability
  • Add to Cart