ENAH11E41518 Enamine

4-(chloromethyl)naphthalene-1-carbonitrile

  • MDL number MFCD18413762
  • Cas Number : 112929-87-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2007591

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2007591
Availability details for
Quantity
  • Availability
  • Add to Cart