ENAH11E4151F Enamine

1-[2-(1,1-difluoroethyl)phenyl]methanamine

  • MDL number MFCD27500639
  • Cas Number : 1638767-86-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2007612

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2007612
Availability details for
Quantity
  • Availability
  • Add to Cart