ENAH11E4152A Enamine

N-(2-aminoethyl)-4-bromo-N-methylaniline

  • MDL number MFCD10031345
  • Cas Number : 851265-79-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2007649

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2007649
Availability details for
Quantity
  • Availability
  • Add to Cart