ENAH11E41531 Enamine

3-tert-butyl-1,2-oxazole-4-carbaldehyde

  • MDL number MFCD31666705
  • Cas Number : 1521395-16-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2007707

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2007707
Availability details for
Quantity
  • Availability
  • Add to Cart