ENAH11E41536 Enamine

2-(3-methylcyclopentyl)ethan-1-amine hydrochloride

  • MDL number MFCD31666738
  • Cas Number : 2228336-68-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2007737

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2007737
Availability details for
Quantity
  • Availability
  • Add to Cart