ENAH11E4153C Enamine

5-(aminomethyl)-5-methylimidazolidine-2,4-dione hydrochloride

  • MDL number MFCD22691110
  • Cas Number : 1009640-21-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2007743

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2007743
Availability details for
Quantity
  • Availability
  • Add to Cart