ENAH11E41541 Enamine

2-methylpropanethioic S-acid

  • MDL number MFCD31690702
  • Cas Number : 44296-44-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2007750

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2007750
Availability details for
Quantity
  • Availability
  • Add to Cart