ENAH11E41544 Enamine

2-methyl-1-benzofuran-7-carbaldehyde

  • MDL number MFCD18822717
  • Cas Number : 89228-67-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2007760

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2007760
Availability details for
Quantity
  • Availability
  • Add to Cart