ENAH11E41545 Enamine

6-amino-4,8-dimethylquinolin-2-ol

  • MDL number MFCD20730363
  • Cas Number : 120689-99-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 286-288
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2007761

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2007761
Availability details for
Quantity
  • Availability
  • Add to Cart