ENAH11E41547 Enamine

4-cyanopiperidine-4-carboxylic acid hydrochloride

  • MDL number MFCD12406600
  • Cas Number : 1205750-37-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2007765

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2007765
Availability details for
Quantity
  • Availability
  • Add to Cart