ENAH11E4154F Enamine

1-[({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)methyl]-3,3-dimethyl-2-oxabicyclo[2.1.1]hexane-4-carboxylic acid

  • MDL number MFCD31666654
  • Cas Number : 2228155-78-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2007917

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2007917
Availability details for
Quantity
  • Availability
  • Add to Cart