ENAH11E41551 Enamine

5-fluoro-1-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione

  • MDL number MFCD20544774
  • Cas Number : 155-16-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2007920

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2007920
Availability details for
Quantity
  • Availability
  • Add to Cart