ENAH11E41553 Enamine

3-[3-(trifluoromethyl)phenyl]-4,5-dihydro-1,2,4-oxadiazol-5-one

  • MDL number MFCD12815752
  • Cas Number : 259271-22-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2007926

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2007926
Availability details for
Quantity
  • Availability
  • Add to Cart