ENAH11E41555 Enamine

4-(aminomethyl)-4-methyl-1lambda6-thiane-1,1-dione hydrochloride

  • MDL number MFCD31666648
  • Cas Number : 2229141-37-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2007949

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2007949
Availability details for
Quantity
  • Availability
  • Add to Cart