ENAH11E41561 Enamine

2-(iodomethyl)-2,3-dihydro-1-benzofuran

  • MDL number MFCD03159146
  • Cas Number : 59152-49-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2007991

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2007991
Availability details for
Quantity
  • Availability
  • Add to Cart