ENAH11E41569 Enamine

1-(6-methoxypyridin-2-yl)ethan-1-ol

  • MDL number MFCD18802369
  • Cas Number : 925417-04-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008042

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008042
Availability details for
Quantity
  • Availability
  • Add to Cart