ENAH11E41571 Enamine

4-bromo-5-chloro-6-methoxypyrimidine

  • MDL number MFCD31379539
  • Cas Number : 2166822-25-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008072

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008072
Availability details for
Quantity
  • Availability
  • Add to Cart