ENAH11E41572 Enamine

4-(2-fluoroacetyl)benzoic acid

  • MDL number MFCD31666524
  • Cas Number : 2062562-03-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008082

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008082
Availability details for
Quantity
  • Availability
  • Add to Cart