ENAH11E41577 Enamine

oxan-3-yl methanesulfonate

  • MDL number MFCD27938761
  • Cas Number : 129888-63-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008094

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008094
Availability details for
Quantity
  • Availability
  • Add to Cart