ENAH11E4157A Enamine

3-benzyl-1-methylpiperazin-2-one hydrochloride

  • MDL number MFCD31666718
  • Cas Number : 2228112-22-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008109

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008109
Availability details for
Quantity
  • Availability
  • Add to Cart