ENAH11E4157B Enamine

methyl 5,5-dioxo-5lambda6-thia-2-azaspiro[3.4]octane-8-carboxylate hydrochloride

  • MDL number MFCD28992063
  • Cas Number : 2177258-03-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008111

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008111
Availability details for
Quantity
  • Availability
  • Add to Cart