ENAH11E41584 Enamine

1-tert-butyl 3-methyl 3-aminoazetidine-1,3-dicarboxylate; trifluoroacetic acid

  • MDL number MFCD30749802
  • Cas Number : 2097953-60-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data CF3COOH
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008127

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008127
Availability details for
Quantity
  • Availability
  • Add to Cart